This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDSN SIRNA Assays ›
Gene Symbol:
Cdsn
Gene Name:
corneodesmosin
Gene Aliases:
AI747712
Chromosome Location:
Chr.17: 35543082 - 35557180 on Build GRCm38
UniGene ID:
Mm.426664
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s118560

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdsn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008424.2 NP_001008424.2 2 371
GenBank mRNA BC055373.1 - 2 368 30058959
BY771914.1 - 2 327

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

single organismal cell-cell adhesion
skin morphogenesis
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline