This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TAS2R130 SIRNA Assays ›
Gene Symbol:
Tas2r130
Gene Name:
taste receptor, type 2, member 130
Gene Aliases:
STC74, T2R30, Tas2r30, Tas2r7, mt2r42
Chromosome Location:
Chr.6: 131629892 - 131630830 on Build GRCm38
UniGene ID:
Mm.377916
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s118641

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tas2r130
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199156.1 NP_954607.1 1 264
GenBank mRNA BC119329.1 - 1 346 8734078

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
signal transducer activity
G-protein coupled receptor activity
bitter taste receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-jgmcd:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0