siRNA Details
| Gene Symbol: Msantd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207277.1 | NP_997160.1 | 1 | 1985 | ||
| GenBank mRNA | AK044284.1 | - | 1 | 1985 | ||
| BC116928.1 | - | 1 | 1053 | 40126247 | ||
| BC116930.1 | - | 1 | 1053 | 40126249 | ||

Msantd1
Genomic Map