This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other E130309D14RIK SIRNA Assays ›
Gene Symbol:
E130309D14Rik
Gene Name:
RIKEN cDNA E130309D14 gene
Gene Aliases:
-
Chromosome Location:
Chr.11: 74619605 - 74641516 on Build GRCm38
UniGene ID:
Mm.274369
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s119141

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: E130309D14Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013784.1 NP_001013806.1 4 1605
GenBank mRNA AK143211.1 - 4 1665
BC150886.1 - 4 1497 9087797

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008767929.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline