siRNA Details
| Gene Symbol: Vmn1r209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013787.1 | NP_001013809.1 | 1 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r209| Gene Symbol: Vmn1r209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013787.1 | NP_001013809.1 | 1 | 527 | ||