This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRR7 SIRNA Assays ›
Gene Symbol:
Prr7
Gene Name:
proline rich 7 (synaptic)
Gene Aliases:
XM_484263
Chromosome Location:
Chr.13: 55464267 - 55473155 on Build GRCm38
UniGene ID:
Mm.315457
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s119218

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prr7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030296.4 NP_001025467.1 3 911
GenBank mRNA AK133129.1 - 3 871
BC138854.1 - 2 579 8861876
BC138856.1 - 2 579 8861878

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_030567, NM_001174101, NM_001174102, BC004261, BC033649, and others.
This siRNA also targets the following transcripts in Rat: NM_001109116, XM_006253652.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0