This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AA467197 SIRNA Assays ›
Gene Symbol:
AA467197
Gene Name:
expressed sequence AA467197
Gene Aliases:
Nmes1, mir-147
Chromosome Location:
Chr.2: 122636971 - 122641087 on Build GRCm38
UniGene ID:
Mm.36783
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s119452

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AA467197
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004174.1 NP_001004174.1 3, 4 209
GenBank mRNA AK141297.1 - 2, 3 312
AK150561.1 - 3, 4 199
BC049633.1 - 3, 4 209 6774154

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-b4tm8:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline