This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KRTAP4-16 SIRNA Assays ›
Gene Symbol:
Krtap4-16
Gene Name:
keratin associated protein 4-16
Gene Aliases:
OTTMUSG00000002196
Chromosome Location:
Chr.11: 99850655 - 99851605 on Build GRCm38
UniGene ID:
Mm.351895
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s119943

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Krtap4-16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013823.1 NP_001013845.1 1 862
GenBank mRNA BC016249.1 - 1 862 4910141

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

aging
hair cycle
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline