This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNHIT3 SIRNA Assays ›
Gene Symbol:
Znhit3
Gene Name:
zinc finger, HIT type 3
Gene Aliases:
Myohd1, Trip3
Chromosome Location:
Chr.11: 84910950 - 84916356 on Build GRCm38
UniGene ID:
Mm.379733
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s120225

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Znhit3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005223.2 NP_001005223.1 4, 5 297
GenBank mRNA AK002888.1 - 4, 5 289
AK003721.1 - 4, 5 296
AK004259.1 - 4, 5 299
AK014350.1 - 4, 5 308
BC089554.1 - 4, 5 385 30300471

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline