This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TEX24 SIRNA Assays ›
Gene Symbol:
Tex24
Gene Name:
testis expressed gene 24
Gene Aliases:
1700108N07Rik, TESF-1
Chromosome Location:
Chr.8: 27344323 - 27349188 on Build GRCm38
UniGene ID:
Mm.84066
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s120314

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tex24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013609.2 NP_001013627.1 1 730
GenBank mRNA AY552600.1 - 1 730
BC139071.1 - 1 699 8862093
BC139072.1 - 1 699 8862094

Back To Top

More Information


Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-lpqbw:80/100.66.77.194:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0