siRNA Details
Gene Symbol: Olfr367-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081010.2 | NP_001074479.2 | 2 | 636 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr367-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081010.2 | NP_001074479.2 | 2 | 636 |