This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM20939 SIRNA Assays ›
Gene Symbol:
Gm20939
Gene Name:
predicted gene, 20939
Gene Aliases:
-
Chromosome Location:
Chr.17: 94864918 - 94878321 on Build GRCm38
UniGene ID:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s120981

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm20939
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024731.2 NP_001019902.2 5 626
GenBank mRNA BC070434.1 - 5 626 6851340

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-nqnmh:80/100.66.77.181:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline