This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM6034 SIRNA Assays ›
Gene Symbol:
Gm6034
Gene Name:
predicted gene 6034
Gene Aliases:
EG547347, H2-gs14-2
Chromosome Location:
Chr.17: 36042961 - 36058645 on Build GRCm38
UniGene ID:
Mm.484641
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s121038

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm6034
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034909.3 NP_001030081.2 3 405
GenBank mRNA AK156795.1 - 406
BC147141.1 - 3 385 9056198
BC147142.1 - 3 385 9056188

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline