siRNA Details
| Gene Symbol: Xlr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045539.2 | NP_001039004.2 | 8 | 754 | ||
| GenBank mRNA | BC051681.1 | - | 8 | 754 | 30021721 | |
| Gene Symbol: Xlr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111293.1 | NP_001104763.1 | 8 | 709 | ||

Xlr5a
Genomic Map