This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC100041034 SIRNA Assays ›
Gene Symbol:
LOC100041034 Sp110
Gene Name:
Sp110 nuclear body protein pseudogene
Sp110 nuclear body protein
Gene Aliases:
-
5031415C07Rik, 52kDa, 5830484A20Rik, ENSMUSG00000075603, Ifi75, Ipr1
Chromosome Location:
Chr.1|NT_166281: 138024 - 163022 on Build GRCm38
Chromosome Location:
Chr.1: 85570741 - 85603891 on Build GRCm38
UniGene ID:
Mm.394746
Mm.335802
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s121231

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100041034
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK163333.1 - 4 1200
Gene Symbol: Sp110
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC099598.1 - 3 1064

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline