This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGS21 SIRNA Assays ›
Gene Symbol:
Rgs21
Gene Name:
regulator of G-protein signalling 21
Gene Aliases:
EG624910
Chromosome Location:
Chr.1: 144519690 - 144567667 on Build GRCm38
UniGene ID:
Mm.389199
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s122034

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rgs21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290269.1 NP_001277198.1 4 579
GenBank mRNA BB635805.1 - 3 504

Back To Top

More Information


Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-c2kgc:80/100.66.76.179:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0