siRNA Details
| Gene Symbol: Gm5114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177890.3 | NP_808558.2 | 2 | 587 | ||
| GenBank mRNA | AK029498.1 | - | 2 | 588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5114| Gene Symbol: Gm5114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177890.3 | NP_808558.2 | 2 | 587 | ||
| GenBank mRNA | AK029498.1 | - | 2 | 588 | ||