siRNA Details
| Gene Symbol: Vmn2r112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104575.1 | NP_001098045.1 | 4 | 1444 | ||
| GenBank mRNA | AB194093.1 | - | 2 | 474 | ||
| AB540943.1 | - | 4 | 1444 | |||
More Information
Set Membership: |
|

Vmn2r112
Genomic Map