siRNA Details
| Gene Symbol: 2610318N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183287.2 | NP_899110.2 | 8 | 1153 | ||
| GenBank mRNA | AK146836.1 | - | 7 | 1045 | ||
| BC039993.1 | - | 8 | 1153 | 1446463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610318N02Rik| Gene Symbol: 2610318N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183287.2 | NP_899110.2 | 8 | 1153 | ||
| GenBank mRNA | AK146836.1 | - | 7 | 1045 | ||
| BC039993.1 | - | 8 | 1153 | 1446463 | ||