siRNA Details
| Gene Symbol: Heatr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045543.2 | NP_001039008.1 | 9 | 950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr9| Gene Symbol: Heatr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045543.2 | NP_001039008.1 | 9 | 950 | ||