siRNA Details
| Gene Symbol: Igkv6-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF045512.1 | - | 1 | 25 | ||
| BC091754.1 | - | 40 | ||||
| X78108.1 | - | 1 | 31 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igkv6-23| Gene Symbol: Igkv6-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF045512.1 | - | 1 | 25 | ||
| BC091754.1 | - | 40 | ||||
| X78108.1 | - | 1 | 31 | |||