This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VMN2R110 SIRNA Assays ›
Gene Symbol:
Vmn2r110
Gene Name:
vomeronasal 2, receptor 110
Gene Aliases:
EG224582
Chromosome Location:
Chr.17: 20573829 - 20596259 on Build GRCm38
UniGene ID:
Mm.469851
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s124161

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vmn2r110
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104572.1 NP_001098042.1 6 1721

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline