This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM6793 SIRNA Assays ›
Gene Symbol:
Gm6793 Hnrnpa3
Gene Name:
heterogeneous nuclear ribonucleoprotein A3 pseudogene
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases:
EG627828
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3
Chromosome Location:
Chr.8: 112013433 - 112015036 on Build GRCm38
Chromosome Location:
Chr.2: 75659230 - 75669407 on Build GRCm38
UniGene ID:
Mm.459502
Mm.316306
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s124768

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm6793
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC157904.1 - 1 332 9088095
BC158074.1 - 1 332 9007371
BC172011.1 - 1 332 9054297
Gene Symbol: Hnrnpa3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053263.1 NP_444493.1 4 363
NM_146130.3 NP_666242.2 4 429
NM_198090.2 NP_932758.1 4 429
GenBank mRNA AB201711.1 - 4 409
AF463524.1 - 4 341
AK010481.1 - 4 357
AK133860.1 - 4 428
AK169663.1 - 4 411
BC023828.1 - 4 382 5343619
BC023908.1 - 4 398 5322033
BC038364.1 - 4 411 5362605
BC057655.1 - 4 342
BC062198.1 - 4 420 2655879
BC064824.1 - 4 394 6477672
BC085474.1 - 3 304 30538435
BC092083.1 - 1 24
BC116250.1 - 4 332 40093600
BC116251.1 - 4 332 40093602
BC157919.1 - 4 332 9007347
BC158038.1 - 4 332 9007341
CJ048391.1 - 4 365
CO045618.1 - 3 213

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: AY363227, AY363226, CB725005.

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
mRNA transport
transmembrane transport
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
RNA transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-swbkf:80/100.66.76.254:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0