siRNA Details
| Gene Symbol: Igkv6-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF045512.1 | - | 2 | 188 | ||
| X78108.1 | - | 2 | 194 | |||
| Gene Symbol: Igkv4-53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M36245.1 | - | 110 | |||

Igkv6-23
Genomic Map