siRNA Details
| Gene Symbol: Myc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012603.2 | NP_036735.2 | 3 | 1501 | ||
| GenBank mRNA | AY679729.1 | - | 3 | 1338 | ||
| AY679730.1 | - | 3 | 1501 | |||
| BC091699.1 | - | 3 | 1307 | 7303974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myc| Gene Symbol: Myc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012603.2 | NP_036735.2 | 3 | 1501 | ||
| GenBank mRNA | AY679729.1 | - | 3 | 1338 | ||
| AY679730.1 | - | 3 | 1501 | |||
| BC091699.1 | - | 3 | 1307 | 7303974 | ||