siRNA Details
| Gene Symbol: Oprd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012617.1 | NP_036749.1 | 2 | 517 | ||
| GenBank mRNA | D16348.1 | - | 2 | 517 | ||
| U00475.1 | - | 2 | 460 | |||
More Information
Other Species Targeted
Set Membership: |
|

Oprd1
Genomic Map