This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AKT2 SIRNA Assays ›
Gene Symbol:
Akt2
Gene Name:
v-akt murine thymoma viral oncogene homolog 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 84400940 - 84451223 on Build Rnor_6.0
UniGene ID:
Rn.87066
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129225

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Akt2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017093.1 NP_058789.1 3 376
XM_008759108.1 XP_008757330.1 4 2775
XM_008759109.1 XP_008757331.1 3 363
XM_008759110.1 XP_008757332.1 3 2640
XM_008759111.1 XP_008757333.1 3 486
GenBank mRNA D30041.1 - 3 376
DQ198085.1 - 2 95

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
glycogen biosynthetic process
glucose metabolic process
regulation of translation
protein phosphorylation
response to osmotic stress
multicellular organism development
insulin receptor signaling pathway
carbohydrate transport
positive regulation of signal transduction
positive regulation of gene expression
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
response to muscle activity
positive regulation of cell migration
positive regulation of vesicle fusion
response to nutrient levels
positive regulation of fatty acid beta-oxidation
peripheral nervous system myelin maintenance
response to insulin
cellular response to insulin stimulus
cellular response to hormone stimulus
negative regulation of RNA splicing
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein kinase B signaling
positive regulation of nitric oxide biosynthetic process
positive regulation of glycogen biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of positive chemotaxis
intracellular protein transmembrane transport
cellular response to organic cyclic compound
protein localization to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of cell motility
positive regulation of glucose import in response to insulin stimulus
protein kinase activity
protein serine/threonine kinase activity
protein kinase C binding
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4v4k2:80/100.66.79.110:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline