This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADRBK1 SIRNA Assays ›
Gene Symbol:
Adrbk1
Gene Name:
adrenergic, beta, receptor kinase 1
Gene Aliases:
BARK1, GRK-2, Grk2
Chromosome Location:
Chr.1: 219536220 - 219544329 on Build Rnor_6.0
UniGene ID:
Rn.13010
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129237

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adrbk1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012776.1 NP_036908.1 16, 17 1638
GenBank mRNA M87854.1 - 16, 17 1638

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001619, X61157, M80776, BC090863, BC037963, and others.
This siRNA also targets the following transcripts in Mouse: NM_130863, AK155691, AF333028, BC053922, BC003196.

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
desensitization of G-protein coupled receptor protein signaling pathway
negative regulation of the force of heart contraction by chemical signal
protein phosphorylation
response to oxidative stress
G-protein coupled receptor signaling pathway
tachykinin receptor signaling pathway
heart development
aging
response to organic cyclic compound
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
receptor internalization
positive regulation of catecholamine secretion
termination of G-protein coupled receptor signaling pathway
response to hydrogen peroxide
follicle-stimulating hormone signaling pathway
negative regulation of G-protein coupled receptor protein signaling pathway
negative regulation of striated muscle contraction
cardiac muscle contraction
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
protein binding
ATP binding
Edg-2 lysophosphatidic acid receptor binding
beta-adrenergic receptor kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline