siRNA Details
| Gene Symbol: Mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012798.1 | NP_036930.1 | 3 | 366 | ||
| GenBank mRNA | BC085755.1 | - | 3 | 327 | 7108924 | |
| U31367.1 | - | 3 | 366 | |||
| X82557.1 | - | 3 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mal| Gene Symbol: Mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012798.1 | NP_036930.1 | 3 | 366 | ||
| GenBank mRNA | BC085755.1 | - | 3 | 327 | 7108924 | |
| U31367.1 | - | 3 | 366 | |||
| X82557.1 | - | 3 | 337 | |||