siRNA Details
| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012936.2 | NP_037068.2 | 5, 6 | 715 | ||
| GenBank mRNA | AF286652.1 | - | 4, 5 | 560 | ||
| BC126096.1 | - | 4, 5 | 588 | 8364407 | ||
| X05900.1 | - | 4, 5 | 421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crybb1| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012936.2 | NP_037068.2 | 5, 6 | 715 | ||
| GenBank mRNA | AF286652.1 | - | 4, 5 | 560 | ||
| BC126096.1 | - | 4, 5 | 588 | 8364407 | ||
| X05900.1 | - | 4, 5 | 421 | |||