siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022507.1 | NP_071952.1 | 6 | 646 | ||
| XM_006239535.2 | XP_006239597.1 | 3 | 3141 | |||
| XM_006239536.2 | XP_006239598.1 | 3 | 2347 | |||
| XM_008764337.1 | XP_008762559.1 | 6 | 547 | |||
| GenBank mRNA | J04532.1 | - | 6 | 646 | ||
| M18332.1 | - | 3 | 199 | |||
| U27191.1 | - | 3 | 411 | |||

Prkcz
Genomic Map