This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CD81 SIRNA Assays ›
Gene Symbol:
Cd81
Gene Name:
Cd81 molecule
Gene Aliases:
Tapa1
Chromosome Location:
Chr.1: 216237697 - 216253311 on Build Rnor_6.0
UniGene ID:
Rn.1975
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130286

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd81
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013087.1 NP_037219.1 7 836
GenBank mRNA FQ229194.1 - 5 416
FQ231991.1 - 7 822
U19894.1 - 7 836

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133655, AK166521, AK170741, AK183391, AK136370, and others.

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
phosphatidylinositol biosynthetic process
protein localization
positive regulation of cell proliferation
response to wounding
positive regulation of B cell proliferation
regulation of growth
regulation of cell proliferation
positive regulation of 1-phosphatidylinositol 4-kinase activity
phosphatidylinositol metabolic process
positive regulation of peptidyl-tyrosine phosphorylation
regulation of cell motility
protein binding
MHC class II protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline