siRNA Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013146.2 | NP_037278.1 | 3, 4 | 753 | ||
| XM_006236260.2 | XP_006236322.1 | 3, 4 | 2303 | |||
| XM_006236261.2 | XP_006236323.1 | 4, 5 | 824 | |||
| XM_006236262.2 | XP_006236324.1 | 4, 5 | 977 | |||

Cald1
Genomic Map