siRNA Details
| Gene Symbol: Htr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013148.1 | NP_037280.1 | 1 | 361 | ||
| GenBank mRNA | L10072.1 | - | 1 | 361 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr5a| Gene Symbol: Htr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013148.1 | NP_037280.1 | 1 | 361 | ||
| GenBank mRNA | L10072.1 | - | 1 | 361 | ||
Set Membership: |
|