siRNA Details
| Gene Symbol: Axin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024355.1 | NP_077331.1 | 8, 9 | 2303 | ||
| GenBank mRNA | AF017757.1 | - | 8, 9 | 2303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Axin2| Gene Symbol: Axin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024355.1 | NP_077331.1 | 8, 9 | 2303 | ||
| GenBank mRNA | AF017757.1 | - | 8, 9 | 2303 | ||