siRNA Details
| Gene Symbol: Cdh22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019161.2 | NP_062034.2 | 3 | 724 | ||
| GenBank mRNA | D83348.1 | - | 3 | 730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh22| Gene Symbol: Cdh22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019161.2 | NP_062034.2 | 3 | 724 | ||
| GenBank mRNA | D83348.1 | - | 3 | 730 | ||