siRNA Details
| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017184.1 | NP_058880.1 | 7 | 560 | ||
| GenBank mRNA | J04993.1 | - | 7 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnni1| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017184.1 | NP_058880.1 | 7 | 560 | ||
| GenBank mRNA | J04993.1 | - | 7 | 560 | ||