siRNA Details
| Gene Symbol: Scn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019265.2 | NP_062138.1 | 9 | 1039 | ||
| XM_008766674.1 | XP_008764896.1 | 9 | 1059 | |||
| GenBank mRNA | AF059030.2 | - | 9 | 1039 | ||
| AJ237852.1 | - | 8 | 988 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn11a| Gene Symbol: Scn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019265.2 | NP_062138.1 | 9 | 1039 | ||
| XM_008766674.1 | XP_008764896.1 | 9 | 1059 | |||
| GenBank mRNA | AF059030.2 | - | 9 | 1039 | ||
| AJ237852.1 | - | 8 | 988 | |||