siRNA Details
| Gene Symbol: Smad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019275.2 | NP_062148.1 | 4 | 590 | ||
| GenBank mRNA | AB010954.1 | - | 4 | 590 | ||
| AF056002.1 | - | 662 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smad4| Gene Symbol: Smad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019275.2 | NP_062148.1 | 4 | 590 | ||
| GenBank mRNA | AB010954.1 | - | 4 | 590 | ||
| AF056002.1 | - | 662 | ||||