This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RESP18 SIRNA Assays ›
Gene Symbol:
Resp18
Gene Name:
regulated endocrine-specific protein 18
Gene Aliases:
-
Chromosome Location:
Chr.9: 82470794 - 82477136 on Build Rnor_6.0
UniGene ID:
Rn.66430
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132233

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Resp18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019278.1 NP_062151.1 2, 3 307
GenBank mRNA BC058147.1 - 2, 3 303 6922403
L25633.1 - 2, 3 307

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-qkfm7:80/100.66.76.162:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0