siRNA Details
| Gene Symbol: Inpp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019311.1 | NP_062184.1 | 2 | 363 | ||
| GenBank mRNA | U55192.1 | - | 2 | 363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inpp5d| Gene Symbol: Inpp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019311.1 | NP_062184.1 | 2 | 363 | ||
| GenBank mRNA | U55192.1 | - | 2 | 363 | ||