siRNA Details
| Gene Symbol: Ywhab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019377.1 | NP_062250.1 | 5 | 793 | ||
| GenBank mRNA | BC076502.1 | - | 5 | 787 | 7100393 | |
| D17446.1 | - | 5 | 788 | |||
| FQ214109.1 | - | 5 | 829 | |||
| FQ229978.1 | - | 5 | 798 | |||
| S55223.1 | - | 5 | 788 | |||
| S83440.1 | - | 5 | 793 | |||

Ywhab
Genomic Map