siRNA Details
| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021694.3 | NP_067726.3 | 26 | 2742 | ||
| GenBank mRNA | AJ236911.1 | - | 24 | 2633 | ||
| BC091218.1 | - | 26 | 2742 | 7107802 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef1| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021694.3 | NP_067726.3 | 26 | 2742 | ||
| GenBank mRNA | AJ236911.1 | - | 24 | 2633 | ||
| BC091218.1 | - | 26 | 2742 | 7107802 | ||