siRNA Details
| Gene Symbol: Epn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033914.1 | NP_001029086.1 | 4 | 1127 | ||
| NM_021852.2 | NP_068624.2 | 3 | 956 | |||
| XM_006246521.2 | XP_006246583.1 | 3 | 875 | |||
| XM_006246522.2 | XP_006246584.1 | 5 | 1209 | |||
| XM_006246523.2 | XP_006246585.1 | 2 | 704 | |||
| GenBank mRNA | AF096269.1 | - | 2 | 697 | ||
| BC094524.1 | - | 4 | 1106 | 7318094 | ||
| FQ211574.1 | - | 5 | 1209 | |||
| FQ211939.1 | - | 4 | 1151 | |||

Epn2
Genomic Map