This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HPSE SIRNA Assays ›
Gene Symbol:
Hpse
Gene Name:
heparanase
Gene Aliases:
Hep, Hpa
Chromosome Location:
Chr.14: 10534358 - 10576686 on Build Rnor_6.0
UniGene ID:
Rn.6392
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134235

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hpse
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022605.1 NP_072127.1 4 611
GenBank mRNA AF184967.1 - 4 611
AF359508.1 - 4 638

Back To Top

More Information


Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
cell adhesion
cell-matrix adhesion
positive regulation of vascular endothelial growth factor production
positive regulation of blood coagulation
heparan sulfate proteoglycan catabolic process
positive regulation of osteoblast proliferation
wound healing
regulation of hair follicle development
positive regulation of hair follicle development
positive regulation of protein kinase B signaling
angiogenesis involved in wound healing
vascular wound healing
heparanase activity
syndecan binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0