siRNA Details
| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022691.1 | NP_073182.1 | 10 | 1526 | ||
| GenBank mRNA | AF032667.1 | - | 10 | 1526 | ||
| FQ231273.1 | - | 1293 | ||||
| Gene Symbol: LOC100911734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768426.1 | XP_008766648.1 | 11 | 1400 | ||
| XM_008768427.1 | XP_008766649.1 | 11 | 1400 | |||
| XM_008768428.1 | XP_008766650.1 | 10 | 1307 | |||
| XM_008768429.1 | XP_008766651.1 | 10 | 1307 | |||
| XM_008768430.1 | XP_008766652.1 | 1568 | ||||

Exoc7
Genomic Map