siRNA Details
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024402.1 | NP_077378.1 | 5 | 1220 | ||
GenBank mRNA | AF008114.1 | - | 5 | 1220 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024402.1 | NP_077378.1 | 5 | 1220 | ||
GenBank mRNA | AF008114.1 | - | 5 | 1220 |