siRNA Details
| Gene Symbol: Gnrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031038.3 | NP_112300.2 | 1, 2 | 609 | ||
| GenBank mRNA | L07646.1 | - | 1, 2 | 595 | ||
| X68980.1 | - | 1, 2 | 559 | |||
| X76635.1 | - | 1, 2 | 548 | |||
More Information
Set Membership: |
|

Gnrhr
Genomic Map