This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MSH2 SIRNA Assays ›
Gene Symbol:
Msh2
Gene Name:
mutS homolog 2
Gene Aliases:
-
Chromosome Location:
Chr.6: 11215951 - 11274916 on Build Rnor_6.0
UniGene ID:
Rn.3174
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135558

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Msh2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031058.1 NP_112320.1 14 2308
GenBank mRNA BC161846.1 - 14 2324 9029023
X93591.1 - 14 2308

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

meiotic mismatch repair
in utero embryonic development
somatic recombination of immunoglobulin genes involved in immune response
oxidative phosphorylation
DNA repair
mismatch repair
postreplication repair
double-strand break repair
meiotic gene conversion
cellular response to DNA damage stimulus
cell cycle arrest
germ cell development
spermatogenesis
determination of adult lifespan
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
response to X-ray
response to UV-B
response to organic cyclic compound
somatic hypermutation of immunoglobulin genes
somatic recombination of immunoglobulin gene segments
B cell mediated immunity
B cell differentiation
intra-S DNA damage checkpoint
response to drug
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
response to amino acid
negative regulation of neuron apoptotic process
maintenance of DNA repeat elements
negative regulation of reciprocal meiotic recombination
isotype switching
negative regulation of DNA recombination
positive regulation of helicase activity
magnesium ion binding
four-way junction DNA binding
Y-form DNA binding
heteroduplex DNA loop binding
double-strand/single-strand DNA junction binding
DNA binding
damaged DNA binding
double-stranded DNA binding
single-stranded DNA binding
ATP binding
protein C-terminus binding
DNA-dependent ATPase activity
ATPase activity
centromeric DNA binding
enzyme binding
protein kinase binding
mismatched DNA binding
guanine/thymine mispair binding
single base insertion or deletion binding
dinucleotide insertion or deletion binding
single guanine insertion binding
single thymine insertion binding
dinucleotide repeat insertion binding
oxidized purine DNA binding
MutLalpha complex binding
protein homodimerization activity
ADP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-vflc8:80/100.66.77.194:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline