siRNA Details
| Gene Symbol: St3gal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031337.2 | NP_112627.2 | 3 | 675 | ||
| XM_006236676.1 | XP_006236738.1 | 2 | 336 | |||
| XM_006236677.1 | XP_006236739.1 | 3 | 478 | |||
| XM_006236679.2 | XP_006236741.1 | 2 | 469 | |||
| XM_006236680.1 | XP_006236742.1 | 2 | 336 | |||
| XM_006236682.1 | XP_006236744.1 | 2 | 275 | |||
| XM_006236684.2 | XP_006236746.1 | 2 | 469 | |||
| XM_008762996.1 | XP_008761218.1 | 2 | 498 | |||
| XM_008762997.1 | XP_008761219.1 | 2 | 498 | |||
| GenBank mRNA | AB018049.1 | - | 2 | 341 | ||
| BC078798.1 | - | 3 | 675 | 7106217 | ||

St3gal5
Genomic Map